Anti-SHPRH Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB34009EA 313 EUR
QEDB34009
Anti-SHPRH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SHPRH (SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR