Anti-RASSF5 Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB34010EA 433 EUR
QEDB34010
Anti-RASSF5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RASSF5 (Ras association (RalGDS/AF-6) domain family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR