Anti-GPC1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB34029EA 313 EUR
QEDB34029
Anti-GPC1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GPC1 (glypican 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR