Anti-CKAP5 Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB34032EA 205 EUR
QEDB34032
Anti-CKAP5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CKAP5 (cytoskeleton associated protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR