Anti-ANO1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB34036EA 433 EUR
QEDB34036
Anti-ANO1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ANO1 (anoctamin 1, calcium activated chloride channel)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR