Anti-HIF1A Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB34039EA 336 EUR
QEDB34039
Anti-HIF1A Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR