Anti-Insulin Growth Factor-1 Receptor (IGF-1R) Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB35101EA 272 EUR
QEDB35101
Anti-Insulin Growth Factor-1 Receptor (IGF-1R) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Insulin Growth Factor-1 Receptor (IGF-1R)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR