Anti-Corticotropin Releasing Factor Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56102EA 369 EUR
QEDB56102
Anti-Corticotropin Releasing Factor Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Corticotropin Releasing Factor
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR