Anti-S100A1 Sheep Polyclonal Antibody

Lieferant: QED Bioscience
QEDB56201EA 230.98 EUR
QEDB56201
Anti-S100A1 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S100A1 (S100 calcium binding protein A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR