Anti-LAMP1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56270EA 0 EUR
QEDB56270
Anti-LAMP1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: LAMP1 (lysosomal-associated membrane protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR