Anti-MAPK14 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56450EA 302 EUR
QEDB56450
Anti-MAPK14 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: MAPK14 (mitogen-activated protein kinase 14)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR