Anti-CD74 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56451EA 302 EUR
QEDB56451
Anti-CD74 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CD74 (CD74 molecule, major histocompatibility complex, class II invariant chain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR