Anti-DLG4 Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB56452EA 349 EUR
QEDB56452
Anti-DLG4 Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DLG4 (discs, large homolog 4 (Drosophila))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR