Anti-CaMKII, phosphorylated Mouse Monoclonal Antibody
Lieferant: QED Bioscience
QEDB56454EA 483 EUR
QEDB56454
Anti-CaMKII, phosphorylated Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CaMKII, phosphorylated
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat