Anti-TXNDC5 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56458EA 355 EUR
QEDB56458
Anti-TXNDC5 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TXNDC5 (thioredoxin domain containing 5 (endoplasmic reticulum))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR