Anti-NR2B Glutamate Receptor Mouse Monoclonal Antibody
Lieferant: QED Bioscience
QEDB56461EA 492 EUR
QEDB56461
Anti-NR2B Glutamate Receptor Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: NR2B Glutamate Receptor
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat