Anti-Shank 1 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56469EA 492 EUR
QEDB56469
Anti-Shank 1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Shank 1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR