Anti-Bassoon Rabbit Polyclonal Antibody

Lieferant: QED Bioscience
QEDB56495EA 482 EUR
QEDB56495
Anti-Bassoon Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Bassoon
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR