Anti-GABRB2 Mouse monoclonal antibody unconjugated

Lieferant: QED Bioscience
QEDB56523EA 355 EUR
QEDB56523
Anti-GABRB2 Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GABRB2 (gamma-aminobutyric acid (GABA) A receptor, beta 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR