Anti-GABRB1 Mouse monoclonal antibody unconjugated
Lieferant: QED Bioscience
QEDB56524EA 492 EUR
QEDB56524
Anti-GABRB1 Mouse monoclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GABRB1 (gamma-aminobutyric acid (GABA) A receptor, beta 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat