Anti-ADAM22 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56527EA 355 EUR
QEDB56527
Anti-ADAM22 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ADAM22 (ADAM metallopeptidase domain 22)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope: extracellular domain
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR