Anti-ADRA2C Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56529EA 355 EUR
QEDB56529
Anti-ADRA2C Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ADRA2C (adrenoceptor alpha 2C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR