Anti-SYT7 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56533EA 355 EUR
QEDB56533
Anti-SYT7 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SYT7 (synaptotagmin VII)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR