Anti-LRP4 (extracellular) Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB56540EA 355 EUR
QEDB56540
Anti-LRP4 (extracellular) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: LRP4 (extracellular)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR