Anti-Amyloid Beta 1-40 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB57002EA 341 EUR
QEDB57002
Anti-Amyloid Beta 1-40 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Amyloid Beta 1-40
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR