Anti-DUX4 Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB57101EA 341 EUR
QEDB57101
Anti-DUX4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DUX4 (double homeobox 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR