Anti-GTF2B Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB70103EA 306 EUR
QEDB70103
Anti-GTF2B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2B (general transcription factor IIB)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR