Anti-PSE1/Ku Protein Mouse Monoclonal Antibody

Lieferant: QED Bioscience
QEDB70105EA 306 EUR
QEDB70105
Anti-PSE1/Ku Protein Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: PSE1/Ku Protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR