Anti-Choline oxidase Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-101-094EA 1020 EUR
ROCK100-101-094
Anti-Choline oxidase Goat Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Choline Oxidase (Alcaligenes species) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Choline oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Alcaligenes
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR