Anti-GST Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-101-200EA 378 EUR
ROCK100-101-200
Anti-GST Goat Polyclonal Antibody
Antikörper
Primary Goat Anti-Glutathion-S-Transferase (Schistosoma japonicum) Reacts with Human

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR