Anti-NAM-DH Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-101-228EA 1020 EUR
ROCK100-101-228
Anti-NAM-DH Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: NAM-DH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Pseudomonas
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR