Anti- Goat Monoclonal Antibody (Unconjugated) [clone: WA-1]

Lieferant: Rockland Immunochemicals
ROCK100-101-230EA 1020 EUR
ROCK100-101-230
Anti- Goat Monoclonal Antibody (Unconjugated) [clone: WA-1]
Antikörper
A working dilution of 1:2,000 to 1:10,000 of the reconstitution concentration is suggested for this product.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: WA-1
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR