Anti-PGMB Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-101-237EA 1950 EUR
ROCK100-101-237
Anti-PGMB Goat Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of b -Phosphoglucomutase (Lactococcus lacti) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PGMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Lactococcus lacti
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR