Anti-ALPI Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-1135EA 1950 EUR
ROCK100-1135
Anti-ALPI Goat Polyclonal Antibody
Antikörper
Primary Goat Anti-ALKALINE PHOSPHATASE (Calf Intestine) Reacts with Pan

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR