Anti-HRP Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-1138EA 239 EUR
ROCK100-1138
Anti-HRP Goat Polyclonal Antibody
Antikörper
Anti-Peroxidase (Horesradish) (Goat) Antibody has been assayed against 1.0 µg of Peroxidase (Horseradish) in a standard ELISA using Alkaline Phosphatase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) and pNPP as a substrate.

Recommended Dilutions: ELISA: 1:9000 - 1:40000; Western Blot: 1:900 - 1:4000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR