Anti-PFKM Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-1156EA 1020 EUR
ROCK100-1156
Anti-PFKM Goat Polyclonal Antibody
Antikörper
This purified antibody has been tested for use in ELISA, immunofluorescence microscopy and western blot.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 48 kDa in size.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: PFKM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR