Anti-LDHA Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-1173EA 1020 EUR
ROCK100-1173
Anti-LDHA Goat Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 36 kDa in size corresponding to LDH western blotting in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:4000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Goat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR