Anti-HRP Guinea Pig Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-2138EA 644 EUR
ROCK100-2138
Anti-HRP Guinea Pig Polyclonal Antibody
Antikörper
Anti-Peroxidase antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Guinea Pig
Isotype:
Reactivity: Pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR