Anti-NFYB Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-101EA 379 EUR
ROCK100-401-101
Anti-NFYB Rabbit Polyclonal Antibody
Antikörper
This product was assayed by immunoblot and found to be reactive against NF-Y (B subunit specific) at a dilution of 1:2,000 followed by reaction with Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat).

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: NFYB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR