Anti-CA2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-136EA 1020 EUR
ROCK100-401-136
Anti-CA2 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-CARBONIC ANHYDRASE II (Human Erythrocytes) Reacts with Pan

Recommended Dilutions: ELISA: 1:115000; Western Blot: 1:2000 - 1:10000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: CA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR