Anti-Luciferase Rabbit Monoclonal Antibody (Unconjugated) [clone: 9B10]

Lieferant: Rockland Immunochemicals
ROCK100-401-137EA 1950 EUR
ROCK100-401-137
Anti-Luciferase Rabbit Monoclonal Antibody (Unconjugated) [clone: 9B10]
Antikörper
Primary Rabbit Anti-LUCIFERASE (Photobacterium fischerii) Reacts with Pan

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Luciferase
Clonality: Monoclonal
Clone: 9B10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Photobacterium fischerii
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR