Anti-TRBOTR Rabbit Monoclonal Antibody (Unconjugated) [clone: 11H10]

Lieferant: Rockland Immunochemicals
ROCK100-401-139EA 1950 EUR
ROCK100-401-139
Anti-TRBOTR Rabbit Monoclonal Antibody (Unconjugated) [clone: 11H10]
Antikörper
Anti-Trypsin Antibody has been assayed against 1.0 µg of Trypsin (Bovine Pancreas) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: TRBOTR
Clonality: Monoclonal
Clone: 11H10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR