Anti-ADA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-140EA 1020 EUR
ROCK100-401-140
Anti-ADA Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-ADENOSINE DEAMINASE (Calf Spleen) Reacts with Pan

  • Recommended Dilutions: ELISA: 1:5000 - 1:10000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
  • Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ADA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR