Anti-EGFR Rabbit Monoclonal Antibody (Unconjugated) [clone: 15A3]

Lieferant: Rockland Immunochemicals
ROCK100-401-149EA 379 EUR
ROCK100-401-149
Anti-EGFR Rabbit Monoclonal Antibody (Unconjugated) [clone: 15A3]
Antikörper
Anti-EGFR antibody is specifically designed for ELISA, immunoblotting and immunoprecipitation.  Reactivity in other assays is likely, but has not been determined.  Recognition of EGFR is independent of the phosphorylation status at tyrosine 1173.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 2.5 µg/ml; Immunoprecipitation: 10 µl;

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: 15A3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR