Anti-CCNB1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-152EA 379 EUR
ROCK100-401-152
Anti-CCNB1 Rabbit Polyclonal Antibody
Antikörper
This antibody is suitable for use in ELISA, immunoblotting, immunoprecipitation and other immuno-logical methods requiring high titer and specificity.   Specific conditions for reactivity and detection of Cyclin B1 should be optimized by the researcher.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR