Anti-CCND1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-153EA 379 EUR
ROCK100-401-153
Anti-CCND1 Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 34 kDa in size corresponding to Cyclin D1 by western blotting.

  • Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR