Anti-CCNT1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-158EA 379 EUR
ROCK100-401-158
Anti-CCNT1 Rabbit Polyclonal Antibody
Antikörper
Suitable for ELISA, immunoprecipitation, immunochemistry, immunoblotting and other immunological methods requiring high titer and specificity.  Anti-Cyclin T1 is suitable for the detection by immunoblot of human, Rat and mouse Cyclin T1.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CCNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR