Anti-CDK4 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-162EA 379 EUR
ROCK100-401-162
Anti-CDK4 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-cdk4 Reacts with Human, Mouse, Rat

Recommended Dilutions: ELISA: 1:500 - 1:2000; Western Blot: 1:200 - 1:500; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR