Anti-CDK9 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-167EA 379 EUR
ROCK100-401-167
Anti-CDK9 Rabbit Polyclonal Antibody
Antikörper
This antibody has been tested for use in ELISA, immunoprecipitation, immunochemistry and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 43 kDa corresponding to CDK9 (PITALRE).

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR