Anti-CDKN1B Rabbit Monoclonal Antibody (Unconjugated) [clone: 5.C3.1]

Lieferant: Rockland Immunochemicals
ROCK100-401-172EA 379 EUR
ROCK100-401-172
Anti-CDKN1B Rabbit Monoclonal Antibody (Unconjugated) [clone: 5.C3.1]
Antikörper
This antibody is suitable for use in ELISA, immunoprecipitation, co-immunoprecipitation and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~27 kDa in size corresponding to p27 by western blotting.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:4000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: 5.C3.1
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR